Gene Transcripts

Gene Symbol
TBK1
Entrez Gene ID
29110
Gene Name
TANK binding kinase 1
Gene Aliases
FTDALS4, NAK, T2K
Chromosome Location
Chr. 12: 64452060 - 64502119 on Build GRCh38
UniGene ID
Hs.505874
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013254.3 NP_037386.1 9 1265
XM_005268809.1 XP_005268866.1 9 1343
XM_005268810.1 XP_005268867.1 9 1325
GenBank mRNA AF174536.1 - 8 1106
AF191838.1 - 9 1168
AK002192.1 - 1 91
AK291039.1 - 9 1210
AK303683.1 - 4 482
BC034950.2 - 9 1205 4824988

View Full Product Details