Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001289480.1 | NP_001276409.1 | 4 | 767 | |
NM_013709.5 | NP_038737.1 | 5 | 881 | ||
GenBank mRNA | AK141199.1 | - | 5 | 882 | |
AK168483.1 | - | 6 | 509 | ||
BC010296.1 | - | 4 | 329 | 3154966 | |
BC058557.1 | - | 4 | 744 | 6313231 | |
BC061488.1 | - | 5 | 821 | 4483634 | |
BC110043.1 | - | 5 | 859 | 5149633 | |
D85926.1 | - | 5 | 364 |