Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_011898.2 | NP_036028.2 | 2 | 769 | |
GenBank mRNA | AB019280.1 | - | 2 | 838 | |
AF176906.1 | - | 2 | 922 | ||
AK037180.1 | - | 2 | 844 | ||
AK048122.1 | - | 2 | 847 | ||
BC050944.1 | - | 2 | 769 | 6821944 | |
BC057005.1 | - | 2 | 769 | 6821944 |