Gene Transcripts

Gene Symbol
Tspyl2
Entrez Gene ID
52808
Gene Name
TSPY-like 2
Gene Aliases
CDA1, CINAP, DENTT, DXBwg1396e, DXHXS1008E, E130307F10Rik
Chromosome Location
Chr. X: 152336852 - 152342506 on Build GRCm38
UniGene ID
Mm.9997
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029836.3 NP_084112.1 4, 5 1279
GenBank mRNA AK018700.1 - 4, 5 1221
AY273809.1 - 4, 5 1120
BC004006.1 - 4, 5 927
BC030334.1 - 2, 3 426
BC043694.1 - 3, 4 1140
BC054393.1 - 4, 5 1199 6491672

View Full Product Details