Gene Transcripts

Gene Symbol
Eif3g
Entrez Gene ID
53356
Gene Name
eukaryotic translation initiation factor 3, subunit G
Gene Aliases
44kDa, D0Jmb4, Eif3s4, TU-189B2, eIF3-p44, p44
Chromosome Location
Chr. 9: 20894349 - 20898590 on Build GRCm38
UniGene ID
Mm.427309
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016876.3 NP_058572.2 7 465
GenBank mRNA AF108214.1 - 7 479
AK038249.1 - 7 475
AK168362.1 - 7 468
BC008511.1 - 7 465 3592537
U70733.1 - 7 455

View Full Product Details