Gene Transcripts

Gene Symbol
Apom
Entrez Gene ID
55938
Gene Name
apolipoprotein M
Gene Aliases
1190010O19Rik, G3a, NG20
Chromosome Location
Chr. 17: 35128997 - 35131752 on Build GRCm38
UniGene ID
Mm.2161
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018816.1 NP_061286.1 3 343
GenBank mRNA AF207820.1 - 3 343
AK004530.1 - 3 388
BC021597.1 - 3 380 5100655

View Full Product Details