Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_019586.3 | NP_062532.2 | 4, 5 | 476 | |
GenBank mRNA | AJ245899.1 | - | 4, 5 | 410 | |
AK003050.1 | - | 4, 5 | 408 | ||
AK031669.1 | - | 4, 5 | 364 | ||
AK149985.1 | - | 4, 5 | 387 | ||
AK160428.1 | - | 4, 5 | 482 | ||
BC024623.1 | - | 4, 5 | 414 | 4217620 | |
BY707310.1 | - | 4, 5 | 476 |