Gene Transcripts

Gene Symbol
Tdh
Entrez Gene ID
58865
Gene Name
L-threonine dehydrogenase
Gene Aliases
Chromosome Location
Chr. 14: 63492347 - 63509092 on Build GRCm38
UniGene ID
Mm.354229
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021480.5 NP_067455.5 9 1399
GenBank mRNA AF134346.2 - 9 1063
AK077571.1 - 9 1074
AY116662.1 - 8 1000
BC058860.1 - 9 1414 30014660

View Full Product Details