Gene Transcripts

Gene Symbol
Dnajc15
Entrez Gene ID
66148
Gene Name
DnaJ (Hsp40) homolog, subfamily C, member 15
Gene Aliases
1110003P16Rik, Dnajd1
Chromosome Location
Chr. 14: 77826217 - 77874917 on Build GRCm38
UniGene ID
Mm.248046
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025384.3 NP_079660.1 3, 4 256
GenBank mRNA AK002598.1 - 3, 4 252
AK003396.1 - 3, 4 266
AK007806.1 - 3, 4 245
AK007833.1 - 3, 4 245
BC027509.1 - 3, 4 256 1364626

View Full Product Details