Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_025671.2 | NP_079947.1 | 6 | 706 | |
GenBank mRNA | AK004916.1 | - | 3 | 1369 | |
AK004943.1 | - | 6 | 707 | ||
AK017497.1 | - | 6 | 707 | ||
AK018534.1 | - | 6 | 707 | ||
AK168946.1 | - | 6 | 624 | ||
BC022762.1 | - | 5 | 554 | ||
BC069872.1 | - | 6 | 709 | 4219293 |