Gene Transcripts

Gene Symbol
Snrnp48
Entrez Gene ID
67797
Gene Name
small nuclear ribonucleoprotein 48 (U11/U12)
Gene Aliases
1110050F08Rik, 6530403A03Rik, C6orf151
Chromosome Location
Chr. 13: 38204911 - 38227663 on Build GRCm38
UniGene ID
Mm.250783
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026382.2 NP_080658.2 4 383
GenBank mRNA AK007431.1 - 4 372
AK012916.1 - 4 378
AK018315.1 - 4 378
AK036713.1 - 4 383
AK087789.1 - 4 378
AK172155.1 - 4 372
BC021443.1 - 4 366 5038605
BC037073.1 - 4 373

View Full Product Details