Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_028177.3 | NP_082453.2 | 2 | 278 | |
GenBank mRNA | AK008788.1 | - | 275 | ||
AK009698.1 | - | 2 | 280 | ||
AK010307.1 | - | 2 | 279 | ||
AK011302.1 | - | 2 | 279 | ||
AK018688.1 | - | 2 | 278 | ||
AK018717.1 | - | 2 | 274 | ||
AK089103.1 | - | 2 | 268 | ||
AK151719.1 | - | 2 | 278 | ||
AK152995.1 | - | 2 | 278 | ||
AK167447.1 | - | 2 | 259 | ||
AK208366.1 | - | 1 | 29 | ||
AK208913.1 | - | 1 | 29 | ||
AK209844.1 | - | 1 | 29 | ||
AK211969.1 | - | 1 | 29 | ||
AK215636.1 | - | 1 | 29 | ||
BC060951.1 | - | 2 | 271 | 6774795 | |
BC092379.1 | - | 2 | 267 | 6815043 |