Gene Transcripts

Gene Symbol
MYL2
Entrez Gene ID
4633
Gene Name
myosin light chain 2
Gene Aliases
CMH10, MLC-2s/v, MLC2
Chromosome Location
Chr. 12: 110910819 - 110920600 on Build GRCh38
UniGene ID
Hs.75535
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000432.3 NP_000423.2 5, 6 424
GenBank mRNA AF020768.1 - 4, 5 358
AK312013.1 - 5, 6 401
BC015821.1 - 5, 6 406 4245364
BC031006.1 - 5, 6 401 4292208
BC031008.1 - 5, 6 403 4297454
CR456962.1 - 4, 5 353
CR541957.1 - 4, 5 353
M22815.1 - 6, 7 372
S69022.1 - 5, 6 430
X14332.1 - 5, 6 350
X57542.1 - 6, 7 372
X66141.1 - 5, 6 383

View Full Product Details