Gene Transcripts

Gene Symbol
Ifitm5
Entrez Gene ID
73835
Gene Name
interferon induced transmembrane protein 5
Gene Aliases
1110003J06Rik, AW213665, Bril, DSPA1, Hrmp1, Hrtm1
Chromosome Location
Chr. 7: 140948962 - 140950239 on Build GRCm38
UniGene ID
Mm.389989
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053088.2 NP_444318.1 2 617
GenBank mRNA AK003363.1 - 2 648
BC103790.1 - 2 617 30287396
FJ200451.1 - 2 669

View Full Product Details