Gene Transcripts

Gene Symbol
Arhgap18
Entrez Gene ID
73910
Gene Name
Rho GTPase activating protein 18
Gene Aliases
4833419J07Rik, AI314906, AI843501
Chromosome Location
Chr. 10: 26695312 - 26918648 on Build GRCm38
UniGene ID
Mm.356496
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_176837.2 NP_789807.1 3 423
GenBank mRNA AK014728.1 - 3 376
AK087800.1 - 3 399
BC030858.1 - 3 423 4194996

View Full Product Details