Gene Transcripts

Gene Symbol
Nhej1
Entrez Gene ID
75570
Gene Name
nonhomologous end-joining factor 1
Gene Aliases
1700029B21Rik, XLF, cernunnos
Chromosome Location
Chr. 1: 74967346 - 75046653 on Build GRCm38
UniGene ID
Mm.442409
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029342.4 NP_083618.3 221
GenBank mRNA AK006481.1 - 215
BC006063.1 - 195 3591824
BC107252.1 - 140 40052353
BC107253.1 - 140 40052354
BC132359.1 - 111 40130636
BC132361.1 - 111 40130638
BY095373.1 - 144

View Full Product Details