Gene Transcripts

Gene Symbol
NEDD9
Entrez Gene ID
4739
Gene Name
neural precursor cell expressed, developmentally down-regulated 9
Gene Aliases
CAS-L, CAS2, CASL, CASS2, HEF1
Chromosome Location
Chr. 6: 11183298 - 11382348 on Build GRCh38
UniGene ID
Hs.37982
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001142393.1 NP_001135865.1 3 705
NM_006403.3 NP_006394.1 2 510
NM_182966.3 NP_892011.2 2 510
NR_073131.1 4 799
GenBank mRNA AK292682.1 - 3 705
BC020686.1 - 2 481
BC040207.1 - 2 496 4820745
BX648041.1 - 4 767
DA660812.1 - 2 511
DA778416.1 - 2 510
L43821.1 - 2 506
U64317.1 - 2 481

View Full Product Details