Gene Transcripts

Gene Symbol
Prss36
Entrez Gene ID
77613
Gene Name
protease, serine 36
Gene Aliases
C330007D15Rik
Chromosome Location
Chr. 7: 127932638 - 127946725 on Build GRCm38
UniGene ID
Mm.334253
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081374.1 NP_001074843.2 15 2701
GenBank mRNA AK021185.1 - 5 657
BC044738.2 - 5 697

View Full Product Details