Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_030207.2 | NP_084483.2 | 17 | 1918 | |
GenBank mRNA | AK046135.1 | - | 12 | 1424 | |
AK046373.1 | - | 12 | 1424 | ||
AK133558.1 | - | 17 | 1866 | ||
BC026390.1 | - | 7 | 655 | ||
BC046305.1 | - | 11 | 1182 | 6493676 | |
BC048950.1 | - | 1 | 42 |