Gene Transcripts

Gene Symbol
NNAT
Entrez Gene ID
4826
Gene Name
neuronatin
Gene Aliases
Peg5
Chromosome Location
Chr. 20: 37521206 - 37523690 on Build GRCh38
UniGene ID
Hs.504703
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001322802.1 NP_001309731.1 3 1132
NM_005386.3 NP_005377.1 3 1137
NM_181689.2 NP_859017.1 2 1056
GenBank mRNA AB002392.1 - 3 1050
BC001768.1 - 3 1081 3353944
U25033.1 - 3 1077
U25034.1 - 2 998

View Full Product Details