Gene Transcripts
- Gene Symbol
- Sytl3
- Entrez Gene ID
- 83672
- Gene Name
- synaptotagmin-like 3
- Gene Aliases
- A430092N21, Slp3, Slp3-a, Slp3-b, mFLJ00307
- Chromosome Location
- Chr. 17: 6658233 - 6738070 on Build GRCm38
- UniGene ID
- Mm.442201
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_031395.2 |
NP_113572.1
|
8, 9 |
1138 |
|
NM_183368.2 |
NP_899224.2
|
2, 3 |
334 |
|
NM_183370.2 |
NP_899226.2
|
6, 7 |
552 |
|
GenBank mRNA |
AB050743.1 |
- |
|
370 |
|
AB057758.1 |
- |
9, 10 |
1138 |
|
AB057764.1 |
- |
10, 11 |
1282 |
|
AB057765.1 |
- |
8, 9 |
1059 |
|
AB057766.1 |
- |
|
480 |
|
AK131174.1 |
- |
8, 9 |
4550 |
|
BC022608.1 |
- |
7, 8 |
761 |
4166796 |
BC106102.1 |
- |
6, 7 |
552 |
3980415 |
BC118936.2 |
- |
9, 10 |
963 |
40091060 |
BC152315.1 |
- |
1, 2 |
768 |
40105882 |
View Full Product Details