Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001159275.1 | NP_001152747.1 | 1 | 610 | |
GenBank mRNA | AF332006.1 | - | 1 | 468 | |
AK077159.1 | - | 1 | 611 | ||
AY583456.1 | - | 1 | 595 | ||
BC114348.1 | - | 1 | 452 | 40051484 |