Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001320653.1 | NP_001307582.1 | 3 | 620 | |
NM_002532.5 | NP_002523.2 | 3 | 620 | ||
XM_011523893.2 | XP_011522195.1 | 4 | 670 | ||
XM_017024694.1 | XP_016880183.1 | 4 | 670 | ||
GenBank mRNA | AK225247.1 | - | 3 | 543 | |
AK298158.1 | - | 4 | 397 | ||
AK299000.1 | - | 3 | 543 | ||
AK308142.1 | - | 3 | 432 | ||
BC000335.2 | - | 3 | 533 | 2822595 | |
BI552947.1 | - | 3 | 563 | ||
Y08612.2 | - | 3 | 579 |