Gene Transcripts

Gene Symbol
Ccl28
Entrez Gene ID
56838
Gene Name
chemokine (C-C motif) ligand 28
Gene Aliases
CCK1, MEC, Scya28
Chromosome Location
Chr. 13: 119610551 - 119654359 on Build GRCm38
UniGene ID
Mm.143745
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020279.3 NP_064675.1 3 386
GenBank mRNA AF220238.1 - 3 332
AK014821.1 - 2 1244
BC040510.1 - 3 398
BC055864.1 - 3 389 4240191

View Full Product Details