Gene Transcripts

Gene Symbol
Ssbp2
Entrez Gene ID
66970
Gene Name
single-stranded DNA binding protein 2
Gene Aliases
1500004K09Rik, 2310079I02Rik, 9330163K02Rik, A830008M03Rik, AU067692, AW558684, Hspc116, SSDP2
Chromosome Location
Chr. 13: 91460979 - 91706175 on Build GRCm38
UniGene ID
Mm.343095
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024186.5 NP_077148.1 2, 3 231
NM_024272.4 NP_077234.2 2, 3 231
GenBank mRNA AK005150.1 - 2, 3 166
AK010220.1 - 2, 3 173
AK013177.1 - 2, 3 157
AK051840.1 - 2, 3 168
AK133699.1 - 2, 3 175
AY037837.1 - 2, 3 187
AY167988.1 - 2, 3 141
BC002176.1 - 2, 3 243 3486988

View Full Product Details