Gene Transcripts

Gene Symbol
TRRAP
Entrez Gene ID
8295
Gene Name
transformation/transcription domain associated protein
Gene Aliases
PAF350/400, PAF400, STAF40, TR-AP, Tra1
Chromosome Location
Chr. 7: 98878490 - 99013243 on Build GRCh38
UniGene ID
Hs.203952
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001244580.1 NP_001231509.1 8 832
NM_003496.3 NP_003487.1 8 832
GenBank mRNA AF076974.1 - 8 832
AF110377.1 - 8 752

View Full Product Details