Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_173495.2 | NP_775766.2 | 3 | 1430 | |
XM_011545449.2 | XP_011543751.1 | 4 | 1515 | ||
GenBank mRNA | AK054858.1 | - | 3 | 1320 | |
AK298796.1 | - | 3 | 1433 | ||
BC121061.2 | - | 3 | 1277 | 40118449 | |
KJ534921.1 | - | 3 | 1422 | ||
KJ535084.1 | - | 4 | 1381 | ||
KJ535090.1 | - | 4 | 1586 | ||
KR270726.1 | - | 2 | 761 |