Gene Transcripts
- Gene Symbol
- NAB1
- Entrez Gene ID
- 4664
- Gene Name
- NGFI-A binding protein 1
- Gene Aliases
- Chromosome Location
- Chr. 2: 190648860 - 190692766 on Build GRCh38
- UniGene ID
- Hs.107474
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001321312.1 |
NP_001308241.1
|
2, 3 |
1137 |
|
NM_001321313.1 |
NP_001308242.1
|
2, 3 |
923 |
|
NM_001321314.1 |
NP_001308243.1
|
1, 2 |
846 |
|
NM_001321315.1 |
NP_001308244.1
|
1, 2 |
846 |
|
NM_005966.3 |
NP_005957.2
|
4, 5 |
1399 |
|
XM_005246582.1 |
XP_005246639.1
|
3, 4 |
1072 |
|
XM_005246583.1 |
XP_005246640.1
|
3, 4 |
1186 |
|
XM_005246586.1 |
XP_005246643.1
|
2, 3 |
879 |
|
XM_005246587.1 |
XP_005246644.1
|
2, 3 |
879 |
|
XM_011511219.2 |
XP_011509521.1
|
3, 4 |
1041 |
|
XM_017004170.1 |
XP_016859659.1
|
3, 4 |
1082 |
|
XM_017004171.1 |
XP_016859660.1
|
2, 3 |
1252 |
|
XM_017004172.1 |
XP_016859661.1
|
2, 3 |
1024 |
|
XM_017004173.1 |
XP_016859662.1
|
3, 4 |
1107 |
|
XM_017004174.1 |
XP_016859663.1
|
2, 3 |
923 |
|
XM_017004175.1 |
XP_016859664.1
|
2, 3 |
1088 |
|
XM_017004176.1 |
XP_016859665.1
|
2, 3 |
991 |
|
GenBank mRNA |
AF045451.1 |
- |
1, 2 |
846 |
|
AF045452.1 |
- |
1, 2 |
846 |
|
AK292446.1 |
- |
2, 3 |
937 |
|
AK296549.1 |
- |
1, 2 |
174 |
|
AK313542.1 |
- |
1, 2 |
944 |
|
BC035724.1 |
- |
4, 5 |
1288 |
5721972 |
U47007.1 |
- |
1, 2 |
846 |
|
View Full Product Details