Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000453.2 | NP_000444.1 | 4 | 867 | |
XM_011528192.2 | XP_011526494.1 | 4 | 1446 | ||
XM_011528193.2 | XP_011526495.1 | 5 | 949 | ||
XM_011528194.2 | XP_011526496.1 | 4 | 354 | ||
XM_017027158.1 | XP_016882647.1 | 5 | 947 | ||
GenBank mRNA | BC105047.1 | - | 4 | 582 | |
BC105049.1 | - | 4 | 582 | 8144052 | |
D87920.1 | - | 4 | 739 | ||
U66088.1 | - | 4 | 867 |