Gene Transcripts

Gene Symbol
PTDSS1
Entrez Gene ID
9791
Gene Name
phosphatidylserine synthase 1
Gene Aliases
LMHD, PSS1, PSSA
Chromosome Location
Chr. 8: 96261886 - 96334552 on Build GRCh38
UniGene ID
Hs.292579
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290225.1 NP_001277154.1 1 318
NM_014754.2 NP_055569.1 1 318
GenBank mRNA AK293036.1 - 1 302
BC002376.2 - 1 291 2961075
BC004192.2 - 1 268 3509418
BC004390.1 - 1 268 3634879
D14694.1 - 1 265
HY087183.1 - 1 318

View Full Product Details