Gene Transcripts
- Gene Symbol
- SNRNP70
- Entrez Gene ID
- 6625
- Gene Name
- small nuclear ribonucleoprotein U1 subunit 70
- Gene Aliases
- RNPU1Z, RPU1, SNRP70, Snp1, U1-70K, U170K, U1AP, U1RNP
- Chromosome Location
- Chr. 19: 49085140 - 49108613 on Build GRCh38
- UniGene ID
- Hs.467097
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001301069.1 |
NP_001287998.1
|
3, 4 |
721 |
|
NM_003089.5 |
NP_003080.2
|
3, 4 |
721 |
|
GenBank mRNA |
AK096783.1 |
- |
3, 4 |
410 |
|
AL560281.3 |
- |
3, 4 |
408 |
|
BC001315.1 |
- |
3, 4 |
394 |
3454479 |
BE397125.1 |
- |
3, 4 |
592 |
|
M22636.1 |
- |
3, 4 |
385 |
|
X06812.1 |
- |
1, 2 |
69 |
|
X06815.1 |
- |
3, 4 |
365 |
|
X84841.1 |
- |
2, 3 |
214 |
|
View Full Product Details