Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001161527.1 | NP_001154999.1 | 5 | 793 | |
NM_001292016.1 | NP_001278945.1 | 4 | 830 | ||
NM_024309.3 | NP_077285.3 | 5 | 1079 | ||
GenBank mRNA | AF372839.1 | - | 5 | 1079 | |
AJ304866.1 | - | 5 | 1026 | ||
AK026176.1 | - | 5 | 792 | ||
AK096296.1 | - | 5 | 1070 | ||
BC002740.2 | - | 5 | 1037 | 3632736 | |
BX386180.2 | - | 4 | 791 | ||
CB157550.1 | - | 1 | 44 |