Gene Transcripts

Gene Symbol
RTP3
Entrez Gene ID
83597
Gene Name
receptor transporter protein 3
Gene Aliases
LTM1, TMEM7, Z3CXXC3
Chromosome Location
Chr. 3: 46497995 - 46500949 on Build GRCh38
UniGene ID
Hs.196584
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031440.1 NP_113628.1 2 481
GenBank mRNA AJ312776.1 - 2 481
AY562237.1 - 2 413
BC131756.1 - 2 442 40119315

View Full Product Details