Gene Transcripts
- Gene Symbol
- NCAPH
- Entrez Gene ID
- 23397
- Gene Name
- non-SMC condensin I complex subunit H
- Gene Aliases
- BRRN1, CAP-H
- Chromosome Location
- Chr. 2: 96335741 - 96377095 on Build GRCh38
- UniGene ID
- Hs.308045
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001281710.1 |
NP_001268639.1
|
2, 3 |
333 |
|
NM_001281711.1 |
NP_001268640.1
|
2, 3 |
294 |
|
NM_015341.4 |
NP_056156.2
|
2, 3 |
366 |
|
XM_005263908.3 |
XP_005263965.1
|
2, 3 |
306 |
|
XM_006712388.3 |
XP_006712451.1
|
2, 3 |
306 |
|
GenBank mRNA |
AK291053.1 |
- |
2, 3 |
307 |
|
AK299248.1 |
- |
2, 3 |
235 |
|
AK308050.1 |
- |
2, 3 |
273 |
|
AK310649.1 |
- |
2, 3 |
234 |
|
BC024211.2 |
- |
2, 3 |
320 |
3504782 |
BI559873.1 |
- |
2, 3 |
354 |
|
BI560881.1 |
- |
2, 3 |
371 |
|
D38553.1 |
- |
2, 3 |
296 |
|
View Full Product Details