Gene Transcripts

Gene Symbol
MTHFSD
Entrez Gene ID
64779
Gene Name
methenyltetrahydrofolate synthetase domain containing
Gene Aliases
Chromosome Location
Chr. 16: 86530176 - 86555235 on Build GRCh38
UniGene ID
Hs.343627
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001159377.1 NP_001152849.1 8 2307
NM_001159378.1 NP_001152850.1 8 2307
NM_001159379.1 NP_001152851.1 8 2304
NM_001159380.1 NP_001152852.1 8 2271
NM_022764.2 NP_073601.2 8 2304
NR_027489.1 9 2377
NR_027490.1 8 2367
XM_005256101.1 XP_005256158.1 8 2347
XM_005256105.1 XP_005256162.1 7 2262
XM_005256106.1 XP_005256163.1 7 2171
XM_006721247.3 XP_006721310.1 7 2178
XM_011523280.2 XP_011521582.1 8 2347
XM_011523285.1 XP_011521587.1 9 2345
XM_011523286.1 XP_011521588.1 9 2445
XM_011523287.1 XP_011521589.1 9 2349
XM_011523288.1 XP_011521590.1 8 2277
XM_011523289.1 XP_011521591.1 8 2354
XM_017023571.1 XP_016879060.1 7 2193
XM_017023572.1 XP_016879061.1 7 2190
XM_017023573.1 XP_016879062.1 7 2170
XM_017023574.1 XP_016879063.1 8 2270
GenBank mRNA AK023060.1 - 8 2282
AK096836.1 - 9 2378
AK125015.1 - 1 2896

View Full Product Details