Gene Transcripts
- Gene Symbol
- Rpl9
- Entrez Gene ID
- 20005
- Gene Name
- ribosomal protein L9
- Gene Aliases
- Chromosome Location
- Chr. 5: 65388364 - 65391431 on Build GRCm38
- UniGene ID
- Mm.300271
- Species
- Mus musculus
- Gene Symbol
- Gm12795
- Entrez Gene ID
- 100415919
- Gene Name
- ribosomal protein L9 pseudogene
- Gene Aliases
- OTTMUSG00000008451
- Chromosome Location
- Chr. : - on Build
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AV373702.1 |
- |
1 |
68 |
|
View Full Product Details