Gene Transcripts

Gene Symbol
Notch2
Entrez Gene ID
18129
Gene Name
notch 2
Gene Aliases
AI853703, N2
Chromosome Location
Chr. 3: 98013538 - 98150367 on Build GRCm38
UniGene ID
Mm.485843
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010928.2 NP_035058.2 16, 17 2768
GenBank mRNA D32210.1 - 16, 17 2762
X68279.1 - 11, 12 1656

View Full Product Details