Gene Transcripts

Gene Symbol
Gpr50
Entrez Gene ID
14765
Gene Name
G-protein-coupled receptor 50
Gene Aliases
H9
Chromosome Location
Chr. X: 71663767 - 71669257 on Build GRCm38
UniGene ID
Mm.33336
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AF065145.1 - 2 455
AK132272.1 - 2 556
BC137982.1 - 2 452 8861001
BC145080.1 - 2 452

View Full Product Details