Gene Transcripts

Gene Symbol
Trim52
Entrez Gene ID
212085
Gene Name
tripartite motif-containing 52
Gene Aliases
4921513B05Rik
Chromosome Location
Chr. 14: 106106198 - 106135862 on Build GRCm38
UniGene ID
Mm.452063
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198601.3 NP_941003.1 1 1433
GenBank mRNA AK029521.1 - 1 1439

View Full Product Details