Gene Transcripts

Gene Symbol
Ero1lb
Entrez Gene ID
67475
Gene Name
ERO1-like beta (S. cerevisiae)
Gene Aliases
1300013B24Rik, 1700065B09Rik, AI447560, ero1-beta
Chromosome Location
Chr. 13: 12565809 - 12609530 on Build GRCm38
UniGene ID
Mm.358706
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026184.2 NP_080460.2 2 363
GenBank mRNA AF489856.1 - 2 169
AK004981.1 - 340
AK006886.1 - 2 226
AK041951.1 - 2 339
BC058721.1 - 2 363 5705507
BC111900.1 - 2 333 40062137
BC112426.1 - 2 333 40062136
BC145888.1 - 2 314 40131191
BC145890.1 - 2 314 40131193

View Full Product Details