Gene Transcripts

Gene Symbol
Foxo3
Entrez Gene ID
56484
Gene Name
forkhead box O3
Gene Aliases
1110048B16Rik, 2010203A17Rik, C76856, FKHRL1, Fkhr2, Foxo3a
Chromosome Location
Chr. 10: 42181845 - 42276746 on Build GRCm38
UniGene ID
Mm.338613
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019740.2 NP_062714.1 3 2060
GenBank mRNA AF114259.1 - 3 2060
AK047413.1 - 3 2018
AK139629.1 - 3 2049

View Full Product Details