Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_006923.3 | NP_008854.2 | 2, 3 | 652 | |
NR_045585.1 | 2, 3 | 349 | |||
GenBank mRNA | BC000500.2 | - | 2, 3 | 367 | |
BC001406.2 | - | 2, 3 | 374 | 3140716 | |
BQ056236.1 | - | 2, 3 | 652 | ||
BT006709.1 | - | 2, 3 | 356 | ||
CD673481.1 | - | 2, 3 | 336 | ||
CR456708.1 | - | 2, 3 | 356 |