Gene Transcripts

Gene Symbol
Rbp2
Entrez Gene ID
24710
Gene Name
retinol binding protein 2, cellular
Gene Aliases
Chromosome Location
Chr. 8: 106503499 - 106527726 on Build Rnor_6.0
UniGene ID
Rn.9828
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012640.2 NP_036772.2 2 182
XM_006243607.1 XP_006243669.1 3 362
GenBank mRNA M13949.1 - 2 182

View Full Product Details