Gene Transcripts

Gene Symbol
Tgfbr3
Entrez Gene ID
29610
Gene Name
transforming growth factor, beta receptor III
Gene Aliases
betaglycan
Chromosome Location
Chr. 14: 3506416 - 3680508 on Build Rnor_6.0
UniGene ID
Rn.9953
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017256.1 NP_058952.1 3 555
GenBank mRNA M77809.1 - 3 555
M80784.1 - 3 540

View Full Product Details