Gene Transcripts
- Gene Symbol
- Tsx
- Entrez Gene ID
- 29391
- Gene Name
- testis specific X-linked gene
- Gene Aliases
- Chromosome Location
- Chr. X: 74269513 - 74283819 on Build Rnor_6.0
- UniGene ID
- Rn.87406
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_019203.1 |
NP_062076.1
|
7 |
633 |
|
GenBank mRNA |
X99797.1 |
- |
7 |
633 |
|
View Full Product Details