Gene Transcripts

Gene Symbol
OR5M3
Entrez Gene ID
219482
Gene Name
olfactory receptor family 5 subfamily M member 3
Gene Aliases
OR11-191
Chromosome Location
Chr. 11: 56469488 - 56470538 on Build GRCh38
UniGene ID
Hs.553631
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004742.2 NP_001004742.2 1 490
GenBank mRNA BC136982.1 - 1 490
BC136983.1 - 1 490 9020975

View Full Product Details