Gene Transcripts

Gene Symbol
KAAG1
Entrez Gene ID
353219
Gene Name
kidney associated antigen 1
Gene Aliases
RU2AS
Chromosome Location
Chr. 6: 24356903 - 24358284 on Build GRCh38
UniGene ID
Hs.697888
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181337.3 NP_851854.1 1 1281
GenBank mRNA AF181722.1 - 1 1281

View Full Product Details