Gene Transcripts

Gene Symbol
MRGPRG-AS1
Entrez Gene ID
283303
Gene Name
MRGPRG antisense RNA 1
Gene Aliases
C11orf36, HSD-40
Chromosome Location
Chr. 11: 3218332 - 3223131 on Build GRCh38
UniGene ID
Hs.151154
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_027138.1 2 690
GenBank mRNA AK093421.1 - 2 690
AK097749.1 - 3 723
AY422795.1 - 1 357
BC104971.1 - 1 395
BC104973.1 - 1 395

View Full Product Details