Gene Transcripts
- Gene Symbol
- Ppp2r1a
- Entrez Gene ID
- 117281
- Gene Name
- protein phosphatase 2, regulatory subunit A, alpha
- Gene Aliases
- PR65
- Chromosome Location
- Chr. 1: 60717386 - 60736629 on Build Rnor_6.0
- UniGene ID
- Rn.4108
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_057140.1 |
NP_476481.1
|
10 |
1170 |
|
GenBank mRNA |
BC083859.1 |
- |
10 |
1170 |
|
View Full Product Details