Gene Transcripts

Gene Symbol
AADAT
Entrez Gene ID
51166
Gene Name
aminoadipate aminotransferase
Gene Aliases
KAT2, KATII, KYAT2
Chromosome Location
Chr. 4: 170060222 - 170091699 on Build GRCh38
UniGene ID
Hs.529735
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286682.1 NP_001273611.1 3 229
NM_001286683.1 NP_001273612.1 3 302
NM_016228.3 NP_057312.1 2 435
NM_182662.1 NP_872603.1 3 217
XM_006714231.2 XP_006714294.1 4 359
GenBank mRNA AF481738.1 - 2 436
AK055952.1 - 3 217
AK314729.1 - 3 304
BC031068.1 - 3 222 5286180

View Full Product Details